Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005326.1 | NP_001005326.1 | 1 | 18 | ||
GenBank mRNA | BC137002.1 | - | 1 | 39 | 9020995 | |
BC137003.1 | - | 1 | 39 | 9020996 |