Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21681
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
7
1599
NM_001256235.1
NP_001243164.1
8
1552
NM_001256236.1
NP_001243165.1
8
1794
NM_001256237.1
NP_001243166.1
7
1690
NM_001256238.1
NP_001243167.1
7
1598
NM_001256239.1
NP_001243168.1
6
1529
NM_001256240.1
NP_001243169.1
6
1541
NM_001283038.1
NP_001269967.1
7
1611
NM_001283039.1
NP_001269968.1
7
1564
NM_001283040.1
NP_001269969.1
5
1183
NM_014489.3
NP_055304.1
7
1724
NR_027016.2
5
1366
NR_027017.2
8
1898
NR_027018.2
8
1563
NR_045923.1
6
1436
NR_045925.1
6
1346
NR_045926.1
6
1358
NR_045927.1
6
1515
NR_045929.1
6
1482
NR_104270.1
6
1424
NR_104271.1
8
1684
NR_104272.1
5
1354
XM_006718181.3
XP_006718244.1
6
2202
XM_006718183.3
XP_006718246.1
8
1619
XM_006718185.2
XP_006718248.1
7
1511
XM_006718186.2
XP_006718249.1
8
1671
XM_006718190.3
XP_006718253.1
7
1827
XM_006718191.3
XP_006718254.1
7
1815
XM_006718192.1
XP_006718255.1
9
1745
XM_006718193.2
XP_006718256.1
8
1552
XM_011519990.2
XP_011518292.2
8
2078
XM_011519991.2
XP_011518293.2
8
2069
XM_011519992.1
XP_011518294.1
9
1850
XM_011519993.2
XP_011518295.2
8
2176
XM_011519994.2
XP_011518296.2
8
2170
XM_011519995.2
XP_011518297.1
8
2001
XM_011519996.1
XP_011518298.1
9
1802
XM_011519997.1
XP_011518299.1
9
2143
XM_011519998.2
XP_011518300.1
9
1821
XM_011519999.1
XP_011518301.1
9
1780
XM_011520000.1
XP_011518302.1
8
1694
XM_011520001.1
XP_011518303.1
7
1486
XM_011520002.1
XP_011518304.1
7
1474
XM_011520003.2
XP_011518305.1
8
2010
XM_011520004.2
XP_011518306.1
8
1685
XM_011520005.2
XP_011518307.1
7
1582
XM_011520006.2
XP_011518308.1
8
1966
XM_011520007.2
XP_011518309.1
9
1735
XM_017017556.1
XP_016873045.1
7
1565
XM_017017557.1
XP_016873046.1
7
1634
XM_017017558.1
XP_016873047.1
7
1808
XM_017017559.1
XP_016873048.1
8
1671
XM_017017560.1
XP_016873049.1
8
1659
GenBank mRNA
AF159615.1
-
7
1726
AL049261.1
-
3
2755
AL096753.1
-
8
1549
BC009930.2
-
7
1487
3506614
BI497274.1
-
1
234
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0