Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SFRP2 SIRNA Assays ›
          Gene Symbol:
          SFRP2
          Gene Name:
          secreted frizzled related protein 2
          Gene Aliases:
          FRP-2, SARP1, SDF-5
          Chromosome Location:
          Chr.4: 153780590 - 153789076 on Build GRCh38
          UniGene ID:
          Hs.481022
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 216821

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SFRP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003013.2 NP_003004.1 3 938
          GenBank mRNA AF311912.1 - 3 934
          AK075372.1 - 3 931
          AK093922.1 - 4 906
          AY359001.1 - 3 740
          BC008666.1 - 3 918 3874262

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          patterning of blood vessels
          chondrocyte development
          outflow tract morphogenesis
          cardiac left ventricle morphogenesis
          apoptotic process
          G-protein coupled receptor signaling pathway
          cell-cell signaling
          response to nutrient
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          male gonad development
          negative regulation of gene expression
          negative regulation of cardiac muscle cell apoptotic process
          negative regulation of epithelial to mesenchymal transition
          positive regulation of endopeptidase activity
          negative regulation of Wnt signaling pathway
          collagen fibril organization
          positive regulation of cell growth
          negative regulation of cell growth
          negative regulation of cell migration
          negative regulation of BMP signaling pathway
          cellular response to extracellular stimulus
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of cell adhesion mediated by integrin
          positive regulation of catenin import into nucleus
          non-canonical Wnt signaling pathway
          post-anal tail morphogenesis
          response to drug
          negative regulation of mesodermal cell fate specification
          embryonic digit morphogenesis
          positive regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of JUN kinase activity
          positive regulation of fat cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract morphogenesis
          negative regulation of epithelial cell proliferation
          negative regulation of peptidyl-tyrosine phosphorylation
          convergent extension involved in axis elongation
          bone morphogenesis
          sclerotome development
          negative regulation of dermatome development
          hematopoietic stem cell proliferation
          cellular response to X-ray
          negative regulation of canonical Wnt signaling pathway
          planar cell polarity pathway involved in neural tube closure
          Wnt signaling pathway involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          regulation of dopaminergic neuron differentiation
          midbrain morphogenesis
          regulation of stem cell division
          negative regulation of planar cell polarity pathway involved in axis elongation
          fibronectin binding
          G-protein coupled receptor activity
          integrin binding
          Wnt-protein binding
          Wnt-activated receptor activity
          receptor agonist activity
          endopeptidase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline