Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CROCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014675.4 | NP_055490.4 | 37 | 6539 | ||
XM_006711058.3 | XP_006711121.1 | 36 | 6330 | |||
XM_011542468.1 | XP_011540770.1 | 37 | 6516 | |||
XM_011542470.2 | XP_011540772.1 | 36 | 6333 | |||
XM_017002913.1 | XP_016858402.1 | 37 | 6495 | |||
XM_017002914.1 | XP_016858403.1 | 35 | 6141 | |||
XM_017002915.1 | XP_016858404.1 | 34 | 6024 | |||
GenBank mRNA | AB007914.2 | - | 35 | 6178 | ||
AF527734.1 | - | 8 | 1738 | |||
DQ139275.1 | - | 24 | 5875 |