Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TBCD SIRNA Assays ›
Gene Symbol:
TBCD
Gene Name:
tubulin folding cofactor D
Gene Aliases:
SSD-1, tfcD
Chromosome Location:
Chr.17: 82752048 - 82943186 on Build GRCh38
UniGene ID:
Hs.464391
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
216917
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TBCD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005993.4
NP_005984.3
24, 25
2231
XM_005256396.4
XP_005256453.1
23, 24
2196
XM_005256399.4
XP_005256456.1
12, 13
1028
XM_005256404.4
XP_005256461.1
12, 13
1070
XM_006722290.2
XP_006722353.1
23, 24
2128
XM_006722291.3
XP_006722354.1
12, 13
1352
XM_006722292.2
XP_006722355.1
10, 11
764
XM_011523589.1
XP_011521891.1
19, 20
1771
XM_011523590.1
XP_011521892.1
19, 20
1852
XM_011523591.1
XP_011521893.1
19, 20
1756
XM_011523593.1
XP_011521895.1
18, 19
1363
XM_011523594.1
XP_011521896.1
11, 12
833
XM_011523595.2
XP_011521897.1
11, 12
911
XM_011523597.2
XP_011521899.1
9, 10
1564
XM_011523598.1
XP_011521900.1
9, 10
622
XM_011523599.2
XP_011521901.1
9, 10
1340
XM_011523600.2
XP_011521902.1
10, 11
784
XM_017024987.1
XP_016880476.1
24, 25
2092
XM_017024989.1
XP_016880478.1
8, 9
478
XM_017024990.1
XP_016880479.1
12, 13
850
GenBank mRNA
AB023205.1
-
24, 25
2210
AJ006417.1
-
24, 25
2143
AK057975.1
-
9, 10
738
AK091959.1
-
23, 24
2117
AK293471.1
-
24, 25
2209
AL096745.1
-
9, 10
759
AL133562.1
-
20, 21
1676
BC003094.1
-
24, 25
2231
3503192
BC006364.1
-
24, 25
2188
4131114
BC039654.1
-
10, 11
584
4865367
BC150142.1
-
8, 9
550
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
tubulin complex assembly
post-chaperonin tubulin folding pathway
negative regulation of cell-substrate adhesion
negative regulation of microtubule polymerization
adherens junction assembly
positive regulation of GTPase activity
bicellular tight junction assembly
GTPase activator activity
protein binding
beta-tubulin binding
chaperone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0