Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIKFYVE SIRNA Assays ›
          Gene Symbol:
          PIKFYVE
          Gene Name:
          phosphoinositide kinase, FYVE-type zinc finger containing
          Gene Aliases:
          CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
          Chromosome Location:
          Chr.2: 208266178 - 208358751 on Build GRCh38
          UniGene ID:
          Hs.744997
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 217007

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PIKFYVE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178000.1 NP_001171471.1 8 1195
          NM_015040.3 NP_055855.2 8 1195
          NM_152671.3 NP_689884.1 6 904
          XM_011510778.2 XP_011509080.1 9 1245
          XM_011510779.2 XP_011509081.1 9 1178
          XM_011510780.2 XP_011509082.1 9 1263
          XM_011510781.2 XP_011509083.1 9 1204
          XM_011510782.2 XP_011509084.1 9 1245
          XM_011510783.2 XP_011509085.1 9 1245
          XM_011510784.2 XP_011509086.1 9 1263
          XM_011510785.2 XP_011509087.1 9 1203
          XM_011510786.2 XP_011509088.1 7 912
          XM_011510787.1 XP_011509089.1 7 921
          XM_011510788.1 XP_011509090.1 6 904
          XM_011510789.2 XP_011509091.1 5 726
          XM_011510790.1 XP_011509092.1 7 1023
          XM_011510792.2 XP_011509094.1 9 1245
          XM_017003568.1 XP_016859057.1 8 1099
          XM_017003569.1 XP_016859058.1 8 1099
          XM_017003570.1 XP_016859059.1 6 930
          XM_017003571.1 XP_016859060.1 5 726
          XM_017003572.1 XP_016859061.1 7 899
          XM_017003573.1 XP_016859062.1 4 607
          XM_017003574.1 XP_016859063.1 3 278
          GenBank mRNA AK127769.1 - 8 1099
          AY457063.1 - 8 1199
          BC017736.2 - 7 1023
          BC032389.1 - 6 904
          BC125052.1 - 7 926
          BC125053.1 - 8 1184
          EU794609.1 - 8 1199

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_006245078, XM_006245072, XM_006245071, XM_006245079, XM_006245077, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011086, AF102777, AK173077, AK165350, AK143565.

          Panther Classification:

          Molecular Function -

          kinase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylinositol biosynthetic process
          receptor-mediated endocytosis
          myelin assembly
          protein localization to nucleus
          intracellular signal transduction
          phosphatidylinositol-3-phosphate biosynthetic process
          retrograde transport, endosome to Golgi
          phosphatidylinositol phosphorylation
          phosphatidylinositol 5-phosphate metabolic process
          regulation of autophagosome assembly
          1-phosphatidylinositol-3-phosphate 5-kinase activity
          protein binding
          ATP binding
          zinc ion binding
          1-phosphatidylinositol-4-phosphate 5-kinase activity
          phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0