Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APBB1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019043.3 | NP_061916.3 | 5 | 704 | ||
XM_006717451.2 | XP_006717514.1 | 4 | 402 | |||
XM_011519514.2 | XP_011517816.1 | 5 | 717 | |||
GenBank mRNA | AB085852.1 | - | 3 | 383 | ||
AK000812.1 | - | 5 | 663 | |||
AK303597.1 | - | 5 | 1264 | |||
AK308916.1 | - | 5 | 685 | |||
AY152730.1 | - | 4 | 470 | |||
BC035636.1 | - | 5 | 616 | |||
BC054516.1 | - | 4 | 482 | 5172946 |