Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARMC9 SIRNA Assays ›
          Gene Symbol:
          ARMC9
          Gene Name:
          armadillo repeat containing 9
          Gene Aliases:
          ARM, KU-MEL-1, NS21
          Chromosome Location:
          Chr.2: 231198546 - 231394991 on Build GRCh38
          UniGene ID:
          Hs.162411
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 217180

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARMC9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271466.2 NP_001258395.1 15, 16 1592
          NM_001291656.1 NP_001278585.1 16, 17 1745
          NM_025139.5 NP_079415.3 16, 17 1747
          XM_011511905.1 XP_011510207.1 16, 17 1665
          XM_011511906.1 XP_011510208.1 16, 17 1665
          XM_011511907.1 XP_011510209.1 15, 16 1566
          XM_011511908.2 XP_011510210.1 16, 17 1665
          XM_011511909.2 XP_011510211.1 16, 17 1665
          XM_011511910.1 XP_011510212.1 16, 17 1665
          XM_011511911.1 XP_011510213.1 16, 17 1665
          XM_011511912.1 XP_011510214.1 16, 17 1665
          XM_011511913.2 XP_011510215.1 16, 17 1665
          XM_011511914.2 XP_011510216.1 16, 17 1665
          XM_011511915.2 XP_011510217.1 16, 17 1665
          XM_011511916.2 XP_011510218.1 16, 17 1665
          XM_011511919.2 XP_011510221.1 6, 7 1053
          XM_017005018.1 XP_016860507.1 16, 17 1923
          XM_017005019.1 XP_016860508.1 16, 17 1841
          XM_017005020.1 XP_016860509.1 16, 17 1923
          XM_017005021.1 XP_016860510.1 16, 17 1665
          XM_017005022.1 XP_016860511.1 16, 17 1665
          XM_017005023.1 XP_016860512.1 16, 17 1923
          XM_017005024.1 XP_016860513.1 15, 16 1566
          XM_017005025.1 XP_016860514.1 16, 17 1665
          XM_017005026.1 XP_016860515.1 16, 17 1864
          GenBank mRNA AB058771.1 - 5, 6 488
          AK022646.1 - 14, 15 1449
          AY219922.1 - 16, 17 1626
          AY929062.1 - 16, 17 1619
          BC004514.2 - 12, 13 1169
          BC065271.1 - 15, 16 1579 6165433
          BX537956.1 - 16, 17 1663

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-bwr8z:80/100.66.76.55:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0