Search
Search

CLEC12B| Gene Symbol: CLEC12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129998.2 | NP_001123470.1 | 4 | 674 | ||
| NM_001319241.1 | NP_001306170.1 | 4 | 666 | |||
| NM_001319242.1 | NP_001306171.1 | 4 | 666 | |||
| NM_205852.3 | NP_995324.2 | 4 | 674 | |||
| NR_135049.1 | 4 | 674 | ||||
| XM_011520658.2 | XP_011518960.1 | 5 | 685 | |||
| XM_017019295.1 | XP_016874784.1 | 5 | 746 | |||
| GenBank mRNA | AK128243.1 | - | 4 | 669 | ||
| AY358810.1 | - | 4 | 674 | |||
| BC036036.1 | - | 4 | 534 | |||
| BC136805.1 | - | 4 | 614 | 9020795 | ||
| BC136806.1 | - | 4 | 614 | 9020796 | ||
| BM563861.1 | - | 533 | ||||
| DQ368812.1 | - | 4 | 541 | |||