Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006602.1 | NP_001006602.1 | 6, 7 | 685 | ||
GenBank mRNA | BC083862.1 | - | 6, 7 | 685 | 7126912 |