Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007122.3 | NP_001007123.1 | 2 | 664 | ||
NM_001281805.1 | NP_001268734.1 | 3 | 781 | |||
NM_001281806.1 | NP_001268735.1 | 2 | 664 | |||
XM_005272425.4 | XP_005272482.1 | 3 | 2407 | |||
XM_011521235.2 | XP_011519537.1 | 3 | 2409 | |||
XM_017021924.1 | XP_016877413.1 | 3 | 2407 | |||
GenBank mRNA | AK122875.1 | - | 2 | 663 | ||
AL833295.1 | - | 2 | 664 | |||
BC130569.1 | - | 2 | 648 | 40146600 | ||
BC144175.1 | - | 3 | 765 | 9052695 | ||
BC144181.1 | - | 2 | 648 | 9052701 |