Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IGF2 SIRNA Assays ›
          Gene Symbol:
          IGF2 INS-IGF2
          Gene Name:
          insulin like growth factor 2
          INS-IGF2 readthrough
          Gene Aliases:
          C11orf43, GRDF, IGF-II, PP9974
          INSIGF
          Chromosome Location:
          Chr.11: 2129112 - 2149603 on Build GRCh38
          Chromosome Location:
          Chr.11: 2129112 - 2161209 on Build GRCh38
          UniGene ID:
          Hs.272259
          Hs.272259
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 217672

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IGF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000612.5 NP_000603.1 4 4693
          NM_001007139.5 NP_001007140.2 5 4667
          NM_001127598.2 NP_001121070.1 5 4380
          NM_001291861.2 NP_001278790.1 4 4044
          NM_001291862.2 NP_001278791.1 4 4215
          GenBank mRNA AF318382.1 - 1 2144
          AK025719.1 - 1 1813
          AY971350.1 - 1 194
          BC011786.2 - 1 211
          BC042127.1 - 2 436
          Gene Symbol: INS-IGF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003512.3 7 4655
          GenBank mRNA BF205917.1 - 1 641

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          ossification
          positive regulation of protein phosphorylation
          platelet degranulation
          glucose metabolic process
          regulation of gene expression by genetic imprinting
          regulation of transcription, DNA-templated
          multicellular organism development
          female pregnancy
          memory
          positive regulation of cell proliferation
          insulin receptor signaling pathway
          response to radiation
          response to nutrient levels
          response to estradiol
          response to nicotine
          insulin receptor signaling pathway via phosphatidylinositol 3-kinase
          wound healing
          positive regulation of activated T cell proliferation
          response to drug
          positive regulation of catalytic activity
          positive regulation of MAPK cascade
          cellular protein metabolic process
          response to ethanol
          positive regulation of glycogen biosynthetic process
          positive regulation of mitotic nuclear division
          negative regulation of natural killer cell mediated cytotoxicity
          positive regulation of insulin receptor signaling pathway
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of cell division
          positive regulation of protein kinase B signaling
          cellular response to mechanical stimulus
          positive regulation of protein serine/threonine kinase activity
          positive regulation of receptor activity
          positive regulation of glycogen (starch) synthase activity
          insulin receptor binding
          insulin-like growth factor receptor binding
          hormone activity
          protein binding
          growth factor activity
          receptor activator activity
          protein serine/threonine kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0