Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdpoz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007222.3 | NP_001007223.2 | 1 | 491 | ||
GenBank mRNA | BC141104.1 | - | 1 | 491 | 9055716 |