Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007796.1 | NP_001007797.1 | 15 | 1883 | ||
XM_006235946.2 | XP_006236008.1 | 14 | 1679 | |||
GenBank mRNA | BC085781.1 | - | 15 | 1926 | 7107433 | |
DQ233644.1 | - | 15 | 1943 | |||
FQ216748.1 | - | 2 | 507 |