Search
Search

Plag1| Gene Symbol: Plag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008316.1 | NP_001008317.1 | 4 | 664 | ||
| XM_008763525.1 | XP_008761747.1 | 5 | 785 | |||
| XM_008763526.1 | XP_008761748.1 | 6 | 3855 | |||
| XM_008763527.1 | XP_008761749.1 | 3 | 5160 | |||
| GenBank mRNA | BC085871.1 | - | 4 | 664 | 7191202 | |