Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS2 SIRNA Assays ›
Gene Symbol:
RIMS2
Gene Name:
regulating synaptic membrane exocytosis 2
Gene Aliases:
OBOE, RAB3IP3, RIM2
Chromosome Location:
Chr.8: 103500026 - 104254430 on Build GRCh38
UniGene ID:
Hs.655271
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21863
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100117.2
NP_001093587.1
4
896
NM_001282881.1
NP_001269810.1
2
501
NM_014677.4
NP_055492.3
2
501
XM_005251106.3
XP_005251163.1
4
896
XM_005251107.3
XP_005251164.1
4
896
XM_011517395.2
XP_011515697.1
8
949
XM_011517398.2
XP_011515700.1
8
949
XM_017014006.1
XP_016869495.1
8
949
XM_017014007.1
XP_016869496.1
8
949
XM_017014008.1
XP_016869497.1
8
949
XM_017014009.1
XP_016869498.1
8
949
XM_017014010.1
XP_016869499.1
8
949
XM_017014011.1
XP_016869500.1
8
949
XM_017014012.1
XP_016869501.1
8
949
XM_017014013.1
XP_016869502.1
7
907
XM_017014014.1
XP_016869503.1
4
896
XM_017014015.1
XP_016869504.1
8
949
XM_017014016.1
XP_016869505.1
8
949
XM_017014017.1
XP_016869506.1
8
949
XM_017014018.1
XP_016869507.1
4
896
XM_017014019.1
XP_016869508.1
6
1028
XM_017014020.1
XP_016869509.1
5
1463
XM_017014021.1
XP_016869510.1
8
949
XM_017014022.1
XP_016869511.1
2
420
XM_017014023.1
XP_016869512.1
8
949
XM_017014024.1
XP_016869513.1
8
949
XM_017014025.1
XP_016869514.1
8
949
XM_017014026.1
XP_016869515.1
6
1169
XM_017014027.1
XP_016869516.1
8
949
XM_017014028.1
XP_016869517.1
8
949
XM_017014029.1
XP_016869518.1
5
1630
XM_017014030.1
XP_016869519.1
4
896
XM_017014031.1
XP_016869520.1
8
949
XM_017014032.1
XP_016869521.1
8
949
XM_017014033.1
XP_016869522.1
7
1749
XM_017014034.1
XP_016869523.1
5
1630
XM_017014035.1
XP_016869524.1
5
1629
XM_017014036.1
XP_016869525.1
2
420
XM_017014037.1
XP_016869526.1
5
1629
XM_017014038.1
XP_016869527.1
4
896
XM_017014039.1
XP_016869528.1
7
1750
XM_017014040.1
XP_016869529.1
2
420
XM_017014041.1
XP_016869530.1
2
420
XM_017014042.1
XP_016869531.1
4
896
XM_017014043.1
XP_016869532.1
2
420
XM_017014044.1
XP_016869533.1
2
420
XM_017014045.1
XP_016869534.1
2
420
XM_017014046.1
XP_016869535.1
2
420
GenBank mRNA
AB018294.1
-
2
429
AK126939.1
-
4
896
AK307790.1
-
2
467
AK308831.1
-
2
301
BC018057.1
-
2
302
BC043144.2
-
2
417
5288275
BP230765.1
-
2
500
CR749374.1
-
2
333
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
positive regulation of gene expression
calcium ion regulated exocytosis
regulation of exocytosis
cAMP-mediated signaling
insulin secretion
regulation of membrane potential
calcium ion-regulated exocytosis of neurotransmitter
spontaneous neurotransmitter secretion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
Rab GTPase binding
ion channel binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0