siRNA Details
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008553.1 | NP_001008553.1 | 9 | 1430 | ||
GenBank mRNA | X80778.1 | - | 9 | 1430 |

Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008553.1 | NP_001008553.1 | 9 | 1430 | ||
GenBank mRNA | X80778.1 | - | 9 | 1430 |