Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008783.2 | NP_001008783.1 | 2 | 1341 | ||
GenBank mRNA | AK295424.1 | - | 2 | 1190 | ||
AK316095.1 | - | 2 | 1341 | |||
BC067217.1 | - | 1 | 574 | |||
BC087842.1 | - | 2 | 1318 | 6191889 |