Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009448.1 | NP_001009448.1 | 3 | 542 | ||
XM_006249653.2 | XP_006249715.1 | 4 | 1180 | |||
GenBank mRNA | AB121032.1 | - | 3 | 542 |