Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SFI1 SIRNA Assays ›
Gene Symbol:
SFI1
Gene Name:
SFI1 centrin binding protein
Gene Aliases:
PISD, PPP1R139, hSfi1p
Chromosome Location:
Chr.22: 31496139 - 31618554 on Build GRCh38
UniGene ID:
Hs.62209
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
219440
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SFI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007467.2
NP_001007468.1
17
2187
NM_001258325.1
NP_001245254.1
15
2022
NM_001258326.1
NP_001245255.1
15
1937
NM_001258327.1
NP_001245256.1
15
1941
NM_014775.3
NP_055590.2
16
2094
XM_005261868.2
XP_005261925.1
17
2185
XM_006724390.2
XP_006724453.1
15
1939
XM_011530570.2
XP_011528872.1
17
2185
XM_011530571.2
XP_011528873.1
17
2181
XM_011530574.2
XP_011528876.1
17
1862
XM_011530575.2
XP_011528877.1
17
2185
XM_011530576.2
XP_011528878.1
16
2113
XM_011530577.2
XP_011528879.1
16
2109
XM_011530579.2
XP_011528881.1
17
1862
XM_011530580.2
XP_011528882.1
16
2092
XM_011530581.2
XP_011528883.1
17
1862
XM_011530582.2
XP_011528884.1
15
1939
XM_011530583.2
XP_011528885.1
15
1935
XM_017029139.1
XP_016884628.1
16
1769
XM_017029140.1
XP_016884629.1
15
2020
XM_017029141.1
XP_016884630.1
16
1769
XM_017029142.1
XP_016884631.1
16
2113
XM_017029143.1
XP_016884632.1
16
2088
XM_017029144.1
XP_016884633.1
16
2092
XM_017029145.1
XP_016884634.1
15
2020
XM_017029147.1
XP_016884636.1
15
2020
XM_017029148.1
XP_016884637.1
15
2016
XM_017029149.1
XP_016884638.1
16
2113
XM_017029150.1
XP_016884639.1
16
2109
XM_017029151.1
XP_016884640.1
16
2088
XM_017029152.1
XP_016884641.1
14
1846
XM_017029153.1
XP_016884642.1
15
1935
XM_017029154.1
XP_016884643.1
15
2016
XM_017029155.1
XP_016884644.1
14
1846
XM_017029156.1
XP_016884645.1
14
1846
XM_017029157.1
XP_016884646.1
13
1348
XM_017029158.1
XP_016884647.1
17
2181
XM_017029160.1
XP_016884649.1
5
371
XM_017029161.1
XP_016884650.1
14
1523
XM_017029162.1
XP_016884651.1
14
1523
GenBank mRNA
AB011114.2
-
16
1958
AK055147.1
-
9
778
AK292408.1
-
17
1763
AK293987.1
-
15
1635
AK303362.1
-
15
1572
BC110814.1
-
15
1538
6500257
BC129926.1
-
13
1480
40124134
BC129945.1
-
16
1867
40124130
DA391957.1
-
3
232
DA534059.1
-
3
177
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
negative regulation of phosphatase activity
protein binding
phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-89mxr:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0