Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SFI1 SIRNA Assays ›
Gene Symbol:
SFI1
Gene Name:
SFI1 centrin binding protein
Gene Aliases:
PISD, PPP1R139, hSfi1p
Chromosome Location:
Chr.22: 31496139 - 31618554 on Build GRCh38
UniGene ID:
Hs.62209
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
219442
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SFI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007467.2
NP_001007468.1
17
2181
NM_001258325.1
NP_001245254.1
15
2016
NM_001258326.1
NP_001245255.1
15
1931
NM_001258327.1
NP_001245256.1
15
1935
NM_014775.3
NP_055590.2
16
2088
XM_005261868.2
XP_005261925.1
17
2179
XM_006724390.2
XP_006724453.1
15
1933
XM_011530570.2
XP_011528872.1
17
2179
XM_011530571.2
XP_011528873.1
17
2175
XM_011530574.2
XP_011528876.1
17
1856
XM_011530575.2
XP_011528877.1
17
2179
XM_011530576.2
XP_011528878.1
16
2107
XM_011530577.2
XP_011528879.1
16
2103
XM_011530579.2
XP_011528881.1
17
1856
XM_011530580.2
XP_011528882.1
16
2086
XM_011530581.2
XP_011528883.1
17
1856
XM_011530582.2
XP_011528884.1
15
1933
XM_011530583.2
XP_011528885.1
15
1929
XM_017029139.1
XP_016884628.1
16
1763
XM_017029140.1
XP_016884629.1
15
2014
XM_017029141.1
XP_016884630.1
16
1763
XM_017029142.1
XP_016884631.1
16
2107
XM_017029143.1
XP_016884632.1
16
2082
XM_017029144.1
XP_016884633.1
16
2086
XM_017029145.1
XP_016884634.1
15
2014
XM_017029147.1
XP_016884636.1
15
2014
XM_017029148.1
XP_016884637.1
15
2010
XM_017029149.1
XP_016884638.1
16
2107
XM_017029150.1
XP_016884639.1
16
2103
XM_017029151.1
XP_016884640.1
16
2082
XM_017029152.1
XP_016884641.1
14
1840
XM_017029153.1
XP_016884642.1
15
1929
XM_017029154.1
XP_016884643.1
15
2010
XM_017029155.1
XP_016884644.1
14
1840
XM_017029156.1
XP_016884645.1
14
1840
XM_017029157.1
XP_016884646.1
13
1342
XM_017029158.1
XP_016884647.1
17
2175
XM_017029160.1
XP_016884649.1
5
365
XM_017029161.1
XP_016884650.1
14
1517
XM_017029162.1
XP_016884651.1
14
1517
GenBank mRNA
AB011114.2
-
16
1952
AK292408.1
-
17
1757
BC110814.1
-
15
1532
6500257
BC129945.1
-
16
1861
40124130
DA391957.1
-
3
226
DA534059.1
-
3
171
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
negative regulation of phosphatase activity
protein binding
phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0