Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYT1L SIRNA Assays ›
Gene Symbol:
MYT1L
Gene Name:
myelin transcription factor 1 like
Gene Aliases:
MRD39, NZF1, ZC2H2C2, ZC2HC4B
Chromosome Location:
Chr.2: 1789113 - 2331388 on Build GRCh38
UniGene ID:
Hs.434418
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
219462
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYT1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303052.1
NP_001289981.1
23
4125
NM_015025.3
NP_055840.2
23
4119
XM_011510318.1
XP_011508620.1
24
4250
XM_011510319.1
XP_011508621.1
24
4360
XM_011510320.2
XP_011508622.1
24
4291
XM_011510321.2
XP_011508623.1
24
4241
XM_011510322.2
XP_011508624.1
24
4054
XM_011510323.1
XP_011508625.1
23
3973
XM_011510324.1
XP_011508626.1
24
4250
XM_011510325.1
XP_011508627.1
24
4244
XM_011510326.1
XP_011508628.1
23
4187
XM_011510327.1
XP_011508629.1
24
4250
XM_011510328.2
XP_011508630.1
24
4250
XM_017003604.1
XP_016859093.1
25
4339
XM_017003605.1
XP_016859094.1
24
3918
XM_017003606.1
XP_016859095.1
24
4354
XM_017003607.1
XP_016859096.1
24
4285
XM_017003609.1
XP_016859098.1
23
4172
XM_017003610.1
XP_016859099.1
24
4271
XM_017003611.1
XP_016859100.1
23
4181
XM_017003612.1
XP_016859101.1
23
4291
XM_017003613.1
XP_016859102.1
23
4291
XM_017003614.1
XP_016859103.1
23
4130
XM_017003615.1
XP_016859104.1
23
4187
XM_017003616.1
XP_016859105.1
23
4187
XM_017003617.1
XP_016859106.1
23
4181
XM_017003618.1
XP_016859107.1
23
4181
XM_017003619.1
XP_016859108.1
23
4291
XM_017003620.1
XP_016859109.1
17
3240
GenBank mRNA
AB029029.2
-
24
3904
AF036943.1
-
23
4020
BC071612.1
-
5
602
BC137272.1
-
23
3922
9021269
BC137273.1
-
23
3922
9021270
BC150281.1
-
24
3904
9007267
BM715930.1
-
3
419
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
nervous system development
cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System