Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPHKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142644.1 | NP_001136116.1 | 7 | 1621 | ||
NM_030623.3 | NP_085126.2 | 7 | 1621 | |||
XM_005246870.4 | XP_005246927.1 | 8 | 2093 | |||
XM_006712777.3 | XP_006712840.1 | 8 | 2054 | |||
XM_006712778.3 | XP_006712841.1 | 8 | 2053 | |||
XM_011511925.2 | XP_011510227.1 | 9 | 2172 | |||
GenBank mRNA | AB051465.1 | - | 1 | 469 | ||
AK127071.1 | - | 7 | 1671 | |||
BC104954.1 | - | 7 | 1621 | 8143957 | ||
BC104956.1 | - | 7 | 1621 | 8143959 | ||
BC143408.1 | - | 7 | 1621 | |||
CR749297.1 | - | 1 | 727 | |||
CR749494.1 | - | 7 | 1603 |