Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANC1 SIRNA Assays ›
Gene Symbol:
TANC1
Gene Name:
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Gene Aliases:
ROLSB, TANC
Chromosome Location:
Chr.2: 158968600 - 159232659 on Build GRCh38
UniGene ID:
Hs.61590
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
219691
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145909.1
NP_001139381.1
27
6406
NM_033394.2
NP_203752.2
27
6425
XM_005246918.4
XP_005246975.1
26
6189
XM_006712810.3
XP_006712873.1
26
6312
XM_006712812.3
XP_006712875.1
26
6313
XM_006712814.3
XP_006712877.1
23
5608
XM_011512048.2
XP_011510350.1
29
6603
XM_011512051.2
XP_011510353.1
28
6468
XM_011512053.2
XP_011510355.1
27
6358
XM_011512055.2
XP_011510357.1
27
6394
XM_011512058.2
XP_011510360.1
25
6100
XM_011512059.2
XP_011510361.1
23
5961
XM_011512062.2
XP_011510364.1
21
5638
XM_017005141.1
XP_016860630.1
26
6217
XM_017005142.1
XP_016860631.1
28
6822
XM_017005143.1
XP_016860632.1
26
6507
XM_017005144.1
XP_016860633.1
26
6196
XM_017005145.1
XP_016860634.1
29
6637
XM_017005146.1
XP_016860635.1
28
6448
XM_017005147.1
XP_016860636.1
27
6397
XM_017005148.1
XP_016860637.1
28
6563
XM_017005149.1
XP_016860638.1
25
6115
XM_017005150.1
XP_016860639.1
25
6097
XM_017005151.1
XP_016860640.1
25
5956
XM_017005152.1
XP_016860641.1
25
5953
XM_017005153.1
XP_016860642.1
24
5902
XM_017005154.1
XP_016860643.1
27
6319
XM_017005155.1
XP_016860644.1
25
6049
XM_017005156.1
XP_016860645.1
23
5638
XM_017005157.1
XP_016860646.1
24
5815
GenBank mRNA
AB051515.1
-
21
5502
AK000401.1
-
4
1180
AK094285.1
-
1
1493
AK128859.1
-
16
4308
AL833258.1
-
1
571
BC037329.1
-
14
3089
BX648238.1
-
2
6507
BX648327.1
-
22
5685
CA394398.1
-
1
384
Back To Top
More Information
Gene Ontology Categories:
Function(s)
myoblast fusion
visual learning
dendritic spine maintenance
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0