Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040611.1 | NP_001035701.1 | 2 | 4505 | ||
NM_130877.2 | NP_570947.2 | 2 | 4505 | |||
GenBank mRNA | AB091827.1 | - | 3 | 4286 | ||
AJ006464.1 | - | 3 | 4352 | |||
BC094558.1 | - | 1 | 1597 |