Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANAPC10 SIRNA Assays ›
Gene Symbol:
ANAPC10
Gene Name:
anaphase promoting complex subunit 10
Gene Aliases:
APC10, DOC1
Chromosome Location:
Chr.4: 144967117 - 145098541 on Build GRCh38
UniGene ID:
Hs.480876
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22107
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANAPC10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256706.1
NP_001243635.1
2, 3
235
NM_001256707.1
NP_001243636.1
3, 4
308
NM_001256708.1
NP_001243637.1
2, 3
441
NM_001256709.1
NP_001243638.1
4, 5
376
NM_001256710.1
NP_001243639.1
2, 3
235
NM_001256711.1
NP_001243640.1
3, 4
308
NM_001256712.1
NP_001243641.1
2, 3
224
NM_001318367.1
NP_001305296.1
2, 3
273
NM_014885.4
NP_055700.2
2, 3
224
XM_011531524.2
XP_011529826.1
3, 4
464
XM_011531525.2
XP_011529827.1
2, 3
251
XM_011531526.2
XP_011529828.1
2, 3
251
XM_011531527.2
XP_011529829.1
2, 3
251
XM_011531529.2
XP_011529831.1
2, 3
251
XM_011531530.2
XP_011529832.1
3, 4
267
XM_011531531.2
XP_011529833.1
2, 3
251
XM_011531532.2
XP_011529834.1
2, 3
225
XM_017007638.1
XP_016863127.1
2, 3
251
XM_017007639.1
XP_016863128.1
3, 4
900
XM_017007640.1
XP_016863129.1
2, 3
292
XM_017007641.1
XP_016863130.1
3, 4
720
XM_017007642.1
XP_016863131.1
3, 4
465
XM_017007643.1
XP_016863132.1
2, 3
251
XM_017007644.1
XP_016863133.1
2, 3
225
GenBank mRNA
AB012109.1
-
2, 3
222
AF132794.1
-
2, 3
179
AL080090.1
-
3, 4
282
AV757106.2
-
2, 3
283
BC005217.1
-
2, 3
201
3929766
BG494966.1
-
4, 5
338
BG530912.1
-
3, 4
279
BI913723.1
-
2, 3
274
BU175327.1
-
2, 3
205
BU567906.1
-
2, 3
444
CN305215.1
-
1, 2
103
CV023091.1
-
1, 2
123
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
mitotic nuclear division
negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
regulation of mitotic metaphase/anaphase transition
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
protein K11-linked ubiquitination
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0