Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014907.2 | NP_055722.2 | 3 | 253 | ||
XM_011517803.2 | XP_011516105.1 | 4 | 655 | |||
XM_011517804.2 | XP_011516106.1 | 3 | 574 | |||
XM_017014480.1 | XP_016869969.1 | 3 | 934 | |||
GenBank mRNA | AB023184.1 | - | 3 | 253 | ||
BC114965.1 | - | 3 | 232 | 40082639 | ||
BX648967.1 | - | 3 | 803 |