Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168647.1 | NP_001162118.1 | 1, 2 | 552 | ||
NM_001168648.1 | NP_001162119.1 | 1, 2 | 552 | |||
NM_001168649.1 | NP_001162120.1 | 1, 2 | 552 | |||
NM_014927.3 | NP_055742.2 | 1, 2 | 552 | |||
XM_011545471.2 | XP_011543773.1 | 1, 2 | 635 | |||
XM_011545472.2 | XP_011543774.1 | 1, 2 | 556 | |||
XM_017029358.1 | XP_016884847.1 | 1, 2 | 556 | |||
GenBank mRNA | AB020709.1 | - | 1, 2 | 528 | ||
AF418269.1 | - | 1, 2 | 108 | |||
AF418270.1 | - | 1, 2 | 108 | |||
AK294728.1 | - | 1, 2 | 514 | |||
BC126121.1 | - | 1, 2 | 107 | 8991837 | ||
BC136289.1 | - | 1, 2 | 107 | 9020276 | ||
BC143839.1 | - | 1, 2 | 107 | 9052354 | ||
BP200041.1 | - | 1, 2 | 555 | |||
DA067208.1 | - | 1, 2 | 552 |