Search
Search

Cad| Gene Symbol: Cad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289522.1 | NP_001276451.1 | 4 | 661 | ||
| NM_001289523.1 | NP_001276452.1 | 4 | 661 | |||
| NM_023525.2 | NP_076014.1 | 4 | 661 | |||
| GenBank mRNA | BC053097.1 | - | 4 | 630 | ||
| BC060119.1 | - | 4 | 628 | |||
| BC137856.1 | - | 4 | 513 | 8860875 | ||
| BC144972.1 | - | 4 | 513 | 9053520 | ||
| BC144973.1 | - | 4 | 513 | |||