Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199065.2 | NP_951020.1 | 1 | 1470 | ||
GenBank mRNA | AK173296.1 | - | 1389 | |||
BC150756.1 | - | 1 | 578 | 9087667 | ||
BC157902.1 | - | 1 | 578 | 9007276 |