Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000695.1 | NP_001000695.1 | 1 | 471 |
Gene Symbol: LOC100910049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748918.1 | XP_003748966.1 | 471 |