Search
Search

CEP350| Gene Symbol: CEP350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014810.4 | NP_055625.4 | 10 | 2211 | ||
| XM_011510191.2 | XP_011508493.1 | 10 | 2170 | |||
| XM_011510198.2 | XP_011508500.1 | 10 | 2170 | |||
| XM_017002929.1 | XP_016858418.1 | 10 | 2170 | |||
| XM_017002930.1 | XP_016858419.1 | 10 | 2208 | |||
| XM_017002931.1 | XP_016858420.1 | 10 | 2170 | |||
| GenBank mRNA | AF287356.1 | - | 10 | 1995 | ||
| AK026986.1 | - | 5 | 935 | |||
| AK095601.1 | - | 10 | 2023 | |||
| BM787594.1 | - | 2 | 597 | |||