Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001938.3 | NP_001001938.1 | 3 | 1620 | ||
NM_001142413.1 | NP_001135885.1 | 3 | 1563 | |||
GenBank mRNA | AK094842.1 | - | 3 | 1561 | ||
AK127854.1 | - | 3 | 1556 | |||
BC136895.1 | - | 3 | 1450 | 9020886 | ||
BC144432.1 | - | 3 | 1452 | 9052953 |