Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006062.2 | NP_006053.2 | 4, 5 | 504 | ||
XM_006711918.3 | XP_006711981.1 | 4, 5 | 591 | |||
XM_017003163.1 | XP_016858652.1 | 3, 4 | 2224 | |||
GenBank mRNA | AK300779.1 | - | 4, 5 | 478 | ||
AK301809.1 | - | 4, 5 | 497 | |||
BC073806.1 | - | 4, 5 | 480 | 6671194 | ||
DC303248.1 | - | 4, 5 | 504 | |||
U50383.1 | - | 4, 5 | 457 |