Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212491.2 | NP_997656.1 | 7 | 739 | ||
GenBank mRNA | BC085934.1 | - | 7 | 739 | 7112671 |