Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ryk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042607.1 | NP_001036072.1 | 5 | 709 | ||
NM_001284258.1 | NP_001271187.1 | 4 | 587 | |||
NM_013649.3 | NP_038677.3 | 5 | 709 | |||
GenBank mRNA | AK144355.1 | - | 4 | 527 | ||
BC032275.1 | - | 5 | 443 | 5370100 | ||
L02210.1 | - | 5 | 684 | |||
L21707.1 | - | 5 | 719 | |||
L38394.1 | - | 5 | 632 | |||
M98547.1 | - | 5 | 673 |