Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP164 SIRNA Assays ›
Gene Symbol:
CEP164
Gene Name:
centrosomal protein 164
Gene Aliases:
NPHP15
Chromosome Location:
Chr.11: 117316346 - 117413266 on Build GRCh38
UniGene ID:
Hs.504009
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22354
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP164
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271933.1
NP_001258862.1
3
215
NM_014956.4
NP_055771.4
4
281
XM_005271453.1
XP_005271510.1
4
552
XM_005271456.1
XP_005271513.1
4
552
XM_005271457.1
XP_005271514.1
4
552
XM_006718788.1
XP_006718851.1
4
552
XM_006718794.1
XP_006718857.1
4
552
XM_011542674.1
XP_011540976.1
4
552
XM_011542690.1
XP_011540992.1
4
552
XM_017017364.1
XP_016872853.1
4
419
XM_017017365.1
XP_016872854.1
4
571
XM_017017366.1
XP_016872855.1
4
354
XM_017017367.1
XP_016872856.1
3
480
XM_017017368.1
XP_016872857.1
4
552
XM_017017369.1
XP_016872858.1
4
552
XM_017017370.1
XP_016872859.1
4
552
XM_017017371.1
XP_016872860.1
4
552
XM_017017372.1
XP_016872861.1
4
552
XM_017017373.1
XP_016872862.1
3
352
XM_017017374.1
XP_016872863.1
3
147
XM_017017375.1
XP_016872864.1
2
276
XM_017017376.1
XP_016872865.1
4
552
XM_017017377.1
XP_016872866.1
4
456
XM_017017378.1
XP_016872867.1
4
552
XM_017017379.1
XP_016872868.1
4
552
XM_017017380.1
XP_016872869.1
3
214
XM_017017381.1
XP_016872870.1
4
552
XM_017017382.1
XP_016872871.1
3
214
XM_017017383.1
XP_016872872.1
3
403
XM_017017384.1
XP_016872873.1
3
202
XM_017017386.1
XP_016872875.1
3
403
GenBank mRNA
AB028975.1
-
3
486
AK302991.1
-
3
255
AK303067.1
-
3
213
BC000602.2
-
3
261
BC054015.1
-
4
554
BX648682.1
-
4
281
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008766182.This siRNA also targets the following transcripts in Mouse: NM_001081373, CK789347, BC076567, AK158671.
Panther Classification:
Molecular Function -
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
DNA repair
mitotic nuclear division
cilium assembly
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-55874ff697-s6fgm:80/10.239.167.51:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0