Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212514.2 | NP_997679.1 | 1, 2 | 138 | ||
XM_006241974.1 | XP_006242036.1 | 2, 3 | 227 | |||
GenBank mRNA | AB162854.1 | - | 1, 2 | 125 | ||
BC098639.1 | - | 1, 2 | 138 | 7324771 |