Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpte | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199257.2 | NP_954866.2 | 20 | 1610 | ||
GenBank mRNA | AJ311311.1 | - | 20 | 1613 | ||
AJ311312.1 | - | 20 | 1698 | |||
AJ311313.1 | - | 20 | 1613 | |||
BC107329.2 | - | 20 | 1405 | 40054415 |