Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sidt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159419.1 | NP_001152891.1 | 26 | 3256 | ||
NM_198034.3 | NP_932151.2 | 25 | 3241 | |||
GenBank mRNA | AK034344.1 | - | 11 | 1406 | ||
AK046836.1 | - | 26 | 3616 | |||
AK171920.1 | - | 25 | 3394 | |||
BC079584.1 | - | 25 | 3241 | 5704672 |