Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137482.1 | - | 1 | 465 | 9021482 | |
BC137483.1 | - | 1 | 465 | 9021483 |