siRNA Details
Gene Symbol: RBBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006910.4 | NP_008841.2 | 7 | 1597 | ||
NM_018703.3 | NP_061173.1 | 7 | 1597 | |||
GenBank mRNA | AB112074.1 | - | 7 | 601 | ||
AB112075.1 | - | 7 | 601 | |||
AF352051.1 | - | 2 | 119 | |||
AK303040.1 | - | 5 | 495 | |||
AY072922.1 | - | 3 | 1346 | |||
BC063524.1 | - | 7 | 664 | |||
BX648661.1 | - | 7 | 537 | |||
X85133.1 | - | 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
