siRNA Details
Gene Symbol: RBBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006910.4 | NP_008841.2 | 7 | 1616 | ||
NM_018703.3 | NP_061173.1 | 7 | 1616 | |||
GenBank mRNA | AB112074.1 | - | 7 | 620 | ||
AB112075.1 | - | 7 | 620 | |||
AF352051.1 | - | 2 | 138 | |||
AK303040.1 | - | 5 | 514 | |||
AY072922.1 | - | 3 | 1365 | |||
BC063524.1 | - | 7 | 683 | |||
BX648661.1 | - | 7 | 556 | |||
X85133.1 | - | 2 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
