Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003529.2 | NP_003520.1 | 1 | 22 | ||
GenBank mRNA | BC066245.1 | - | 1 | 22 | 7002068 | |
BC066246.1 | - | 1 | 22 | 7002069 | ||
BC066247.1 | - | 1 | 22 | 7002070 | ||
BC067490.1 | - | 1 | 22 | 7002071 | ||
BC067491.1 | - | 1 | 22 | 7002072 | ||
BC069303.1 | - | 1 | 22 | 7262697 | ||
CR541858.1 | - | 1 | 22 |