Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282550.1 | NP_001269479.1 | 2 | 301 | ||
NM_001282551.1 | NP_001269480.1 | 2 | 397 | |||
NM_001282552.2 | NP_001269481.1 | 2 | 482 | |||
NM_001282554.1 | NP_001269483.1 | 2 | 397 | |||
NM_014962.3 | NP_055777.1 | 1 | 719 | |||
NM_181443.3 | NP_852108.2 | 2 | 482 | |||
XM_017027728.1 | XP_016883217.1 | 3 | 630 | |||
XM_017027729.1 | XP_016883218.1 | 3 | 562 | |||
XM_017027730.1 | XP_016883219.1 | 1 | 741 | |||
GenBank mRNA | AB023169.1 | - | 1 | 667 | ||
AK299281.1 | - | 2 | 482 | |||
AK300371.1 | - | 1 | 93 | |||
AK316256.1 | - | 2 | 373 | |||
BC093677.1 | - | 1 | 190 | |||
BC101527.1 | - | 1 | 189 | 8069033 | ||
BC109315.1 | - | 2 | 330 | 40036410 | ||
BC109316.1 | - | 2 | 330 | 40036411 | ||
CB961606.1 | - | 2 | 403 | |||
DC314595.1 | - | 2 | 397 | |||
DC418438.1 | - | 2 | 301 | |||
DR759697.1 | - | 2 | 337 |