siRNA Details
Gene Symbol: DNAJB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005494.2 | NP_005485.1 | 7 | 717 | ||
NM_058246.3 | NP_490647.1 | 7 | 716 | |||
XM_005249515.2 | XP_005249572.1 | 7 | 708 | |||
XM_005249516.2 | XP_005249573.1 | 7 | 852 | |||
XM_006715823.1 | XP_006715886.1 | 7 | 708 | |||
XM_011515704.1 | XP_011514006.1 | 7 | 687 | |||
GenBank mRNA | AB014888.1 | - | 7 | 662 | ||
AB015798.1 | - | 7 | 709 | |||
AB015799.1 | - | 7 | 709 | |||
AB209859.1 | - | 7 | 710 | |||
AF060703.1 | - | 7 | 707 | |||
AF075601.1 | - | 7 | 694 | |||
AK124827.1 | - | 2 | 1739 | |||
AK223601.1 | - | 7 | 727 | |||
AK291953.1 | - | 7 | 699 | |||
AK293025.1 | - | 7 | 700 | |||
AK297796.1 | - | 6 | 607 | |||
AK297989.1 | - | 7 | 576 | |||
AK301152.1 | - | 5 | 555 | |||
AL136707.1 | - | 7 | 721 | |||
AL832124.1 | - | 2 | 4333 | |||
BC000177.2 | - | 7 | 708 | 2900421 | ||
BC002446.2 | - | 7 | 716 | 3346070 | ||
BC043270.1 | - | 2 | 2428 | |||
CR533498.1 | - | 6 | 562 |
Gene Symbol: TMEM135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF080569.1 | - | 7 | 617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
