siRNA Details
Gene Symbol: DNAJB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058246.3 | NP_490647.1 | 9, 10 | 1057 | ||
XM_017011633.1 | XP_016867122.1 | 6, 7 | 725 | |||
GenBank mRNA | AB015798.1 | - | 9, 10 | 1050 | ||
AK223601.1 | - | 9, 10 | 1068 | |||
AK297497.1 | - | 6, 7 | 704 | |||
AK297796.1 | - | 8, 9 | 948 | |||
AK297989.1 | - | 9, 10 | 917 | |||
AK301152.1 | - | 7, 8 | 896 | |||
BC002446.2 | - | 9, 10 | 1057 | 3346070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
