Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004483.4 | NP_004474.2 | 5 | 690 | ||
NR_033249.1 | 4 | 626 | ||||
XM_017023136.1 | XP_016878625.1 | 5 | 707 | |||
XM_017023137.1 | XP_016878626.1 | 5 | 650 | |||
GenBank mRNA | BC000790.1 | - | 1 | 614 | 3451361 | |
BC009065.1 | - | 1 | 649 | |||
BC020922.1 | - | 1 | 653 | 4247157 | ||
BF798008.1 | - | 5 | 561 | |||
BG708388.1 | - | 5 | 723 | |||
CN283805.1 | - | 4 | 514 | |||
M69175.1 | - | 5 | 617 |
Gene Symbol: GCSHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033248.1 | 1 | 534 | |||
GenBank mRNA | AK092810.1 | - | 2 | 2418 | ||
AV703937.1 | - | 1 | 662 | |||
BM685716.1 | - | 1 | 287 |