Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SETX SIRNA Assays ›
          Gene Symbol:
          SETX
          Gene Name:
          senataxin
          Gene Aliases:
          ALS4, AOA2, SCAR1, bA479K20.2
          Chromosome Location:
          Chr.9: 132261440 - 132356726 on Build GRCh38
          UniGene ID:
          Hs.460317
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 22481

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SETX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015046.5 NP_055861.3 16 6356
          XM_005272171.1 XP_005272228.1 16 6357
          XM_005272172.2 XP_005272229.1 17 6455
          XM_005272173.2 XP_005272230.1 16 6493
          XM_011518404.2 XP_011516706.1 17 6497
          XM_011518405.2 XP_011516707.1 16 6515
          XM_011518406.2 XP_011516708.1 16 6357
          XM_011518407.1 XP_011516709.1 16 6357
          XM_011518408.2 XP_011516710.1 16 6357
          XM_017014495.1 XP_016869984.1 16 6428
          XM_017014496.1 XP_016869985.1 7 1024
          XM_017014497.1 XP_016869986.1 7 1393
          GenBank mRNA AB014525.2 - 14 6133
          AK001456.1 - 6 892
          AK226007.1 - 7 942
          AY362728.1 - 14 6180
          BC032600.2 - 7 933
          BC032622.1 - 7 1234
          BC078166.1 - 6 893
          BC106017.1 - 5 744
          BC137350.1 - 15 6290 9021348
          BX537849.1 - 16 6341
          BX538166.1 - 16 6359
          CR749249.1 - 7 4837
          KU178559.1 - 4 626
          KU178560.1 - 4 626

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          double-strand break repair
          DNA recombination
          DNA-templated transcription, termination
          termination of RNA polymerase II transcription
          mRNA splice site selection
          RNA processing
          cellular response to DNA damage stimulus
          spermatogenesis
          circadian rhythm
          fibroblast growth factor receptor signaling pathway
          positive regulation of neuron projection development
          DNA duplex unwinding
          positive regulation of RNA splicing
          cellular response to oxidative stress
          negative regulation of apoptotic process
          protein kinase B signaling
          cellular response to fibroblast growth factor stimulus
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of DNA-templated transcription, termination
          cellular response to hydrogen peroxide
          cellular response to retinoic acid
          positive regulation of DNA-templated transcription, initiation
          positive regulation of termination of RNA polymerase II transcription, poly(A)-coupled
          transcription termination site sequence-specific DNA binding
          DNA binding
          DNA helicase activity
          protein binding
          ATP binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0