Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYCBP2 SIRNA Assays ›
Gene Symbol:
MYCBP2
Gene Name:
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases:
Myc-bp2, PAM, Phr
Chromosome Location:
Chr.13: 77044657 - 77327066 on Build GRCh38
UniGene ID:
Hs.591221
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22486
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYCBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015057.4
NP_055872.4
3
767
XM_005266299.2
XP_005266356.1
3
791
XM_006719779.2
XP_006719842.1
3
791
XM_006719780.2
XP_006719843.1
3
791
XM_006719781.2
XP_006719844.1
3
791
XM_006719782.2
XP_006719845.1
3
791
XM_006719783.2
XP_006719846.1
3
791
XM_006719784.2
XP_006719847.1
3
791
XM_006719785.2
XP_006719848.1
3
791
XM_006719786.2
XP_006719849.1
3
791
XM_006719787.2
XP_006719850.1
3
791
XM_006719788.2
XP_006719851.1
3
791
XM_006719789.2
XP_006719852.1
3
791
XM_011535004.2
XP_011533306.1
2
330
XM_017020454.1
XP_016875943.1
3
791
XM_017020455.1
XP_016875944.1
3
791
XM_017020456.1
XP_016875945.1
3
791
XM_017020457.1
XP_016875946.1
3
791
XM_017020458.1
XP_016875947.1
3
791
XM_017020459.1
XP_016875948.1
3
791
XM_017020460.1
XP_016875949.1
3
791
XM_017020461.1
XP_016875950.1
3
791
XM_017020462.1
XP_016875951.1
3
791
XM_017020463.1
XP_016875952.1
3
791
XM_017020464.1
XP_016875953.1
3
791
XM_017020465.1
XP_016875954.1
3
791
XM_017020466.1
XP_016875955.1
3
791
XM_017020467.1
XP_016875956.1
3
791
XM_017020468.1
XP_016875957.1
2
278
GenBank mRNA
AF075587.1
-
4
532
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
protein ubiquitination
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0